Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH62234MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62234MD 250 mg
AAH6223403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62234MD Supplier Thermo Scientific Chemicals Supplier No. H62234MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1887126-77-3 | Molecular Formula: C12H16BFO3 | Molecular Weight: 238.065 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1887126-77-3
Molecular Formula C12H16BFO3
Molecular Weight (g/mol) 238.065
InChI Key KBGOBMOAZQOCTA-UHFFFAOYSA-N
Synonym 5-Fluoro-3-hydroxyphenylboronic acid pinacol ester
PubChem CID 72219551
IUPAC Name 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)O

Specifications

Quantity 250 mg
Formula Weight 238.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.