accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl

CAS RN 1888-71-7

IUPAC Name: hexachloroprop-1-ene
Synonyms: hexachloropropene
Molecular Weight (g/mol): 248.73
Molecular Formula: C3Cl6
InChi Key: VFDYKPARTDCDCU-UHFFFAOYSA-N
SMILES: ClC(Cl)=C(Cl)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results