accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBrO

CAS RN 188813-05-0

IUPAC Name: 3-bromo-5-chlorobenzaldehyde
Synonyms: 3-bromo-5-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: JGMGDYUVFBBCEQ-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(C=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results