accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNOOHCH3H3CCH3

CAS RN 188844-52-2

IUPAC Name: 4-{14,14,17,17-tetramethyl-2-thia-9-azatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(19),3,5,7,9,11,13(18)-heptaen-10-yl}benzoic acid
Synonyms: 4-{14,14,17,17-tetramethyl-2-thia-9-azatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(19),3,5,7,9,11,13(18)-heptaen-10-yl}benzoic acid
Molecular Weight (g/mol): 441.59
Molecular Formula: C28H27NO2S
InChi Key: PFGCWQPTOKPRRK-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=C1SC3=CC=CC=C3N=C(C3=CC=C(C=C3)C(O)=O)C1=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results