accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NCH3O(R)HNO(S)OHOHNONHO(S)NH2NH2NNHO

CAS RN 188968-51-6

IUPAC Name: 2-[(2S,5R,8S,11S)-5-benzyl-11-{3-[(diaminomethylidene)amino]propyl}-7-methyl-3,6,9,12,15-pentaoxo-8-(propan-2-yl)-1,4,7,10,13-pentaazacyclopentadecan-2-yl]acetic acid
Synonyms: cilengitide
Molecular Weight (g/mol): 588.67
Molecular Formula: C27H40N8O7
InChi Key: AMLYAMJWYAIXIA-VWNVYAMZSA-N
SMILES: CC(C)[C@@H]1N(C)C(=O)[C@@H](CC2=CC=CC=C2)NC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@H](CCCN=C(N)N)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)