accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOFFF

CAS RN 1891-90-3

IUPAC Name: 4-(trifluoromethyl)benzamide
Synonyms: P-trifluoromethylbenzamide
Molecular Weight (g/mol): 189.14
Molecular Formula: C8H6F3NO
InChi Key: WEJHBEDHLLBJFW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results