Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)benzamide 98.0+%, TCI America™
SDP

Catalog No. T34745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34745G 5 g
T347425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34745G Supplier TCI America Supplier No. T34745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1891-90-3
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD00007998
InChI Key WEJHBEDHLLBJFW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzamide, p-trifluoromethylbenzamide, p-trifluoromethyl benzamide, benzamide, 4-trifluoromethyl, pubchem5482, acmc-20a3xk, 4-carbamoylbenzotrifluoride, 4-trifluoromethyl-benzamide, 4-cf3-c6h4conh2, n-4-trifluoromethyl benzamide
PubChem CID 74684
IUPAC Name 4-(trifluoromethyl)benzamide
SMILES NC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 189.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.