accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHH2N

CAS RN 1892-22-4

IUPAC Name: 3-aminopiperidin-2-one
Synonyms: 3-amino-2-piperidone
Molecular Weight (g/mol): 114.15
Molecular Formula: C5H10N2O
InChi Key: YCCMTCQQDULIFE-UHFFFAOYNA-N
SMILES: NC1CCCNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results