Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-piperidone 98.0+%, TCI America™
SDP

Catalog No. A24175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24175G 5 g
A24171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24175G Supplier TCI America Supplier No. A24175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1892-22-4
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD06809610,MFCD09258624
InChI Key YCCMTCQQDULIFE-UHFFFAOYNA-N
Synonym 2-Amino-delta-valerolactam
PubChem CID 5200225
ChEBI CHEBI:76341
IUPAC Name 3-aminopiperidin-2-one
SMILES NC1CCCNC1=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 114.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-piperidone
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.