accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18928-91-1

IUPAC Name: 1,3-diethyl 2-cyclopentylpropanedioate
Synonyms: 1,3-diethyl 2-cyclopentylpropanedioate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: WXPOKLNJWFXXQO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C1CCCC1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results