Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Cyclopentylmalonate 95.0+%, TCI America™
SDP

Catalog No. C094425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C094425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C094425ML Supplier TCI America Supplier No. C094425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18928-91-1
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.288
MDL Number MFCD00059514
InChI Key WXPOKLNJWFXXQO-UHFFFAOYSA-N
Synonym Cyclopentylmalonic Acid Diethyl Ester
PubChem CID 249269
IUPAC Name diethyl 2-cyclopentylpropanedioate
SMILES CCOC(=O)C(C1CCCC1)C(=O)OCC

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
Formula Weight 228.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Cyclopentylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.