accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFFCl(Z)

CAS RN 18931-60-7

IUPAC Name: (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one
Synonyms: (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one
Molecular Weight (g/mol): 250.6
Molecular Formula: C10H6ClF3O2
InChi Key: RESLPNKXHQNERF-YVMONPNESA-N
SMILES: O\C(=C/C(=O)C(F)(F)F)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results