Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98.0+%, TCI America™
SDP

Catalog No. C31395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31395G 5 g
C31391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31395G Supplier TCI America Supplier No. C31395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18931-60-7
Molecular Formula C10H6ClF3O2
Molecular Weight (g/mol) 250.60
MDL Number MFCD00511276
InChI Key RESLPNKXHQNERF-YVMONPNESA-N
PubChem CID 2773832
IUPAC Name (3Z)-4-(4-chlorophenyl)-1,1,1-trifluoro-4-hydroxybut-3-en-2-one
SMILES O\C(=C/C(=O)C(F)(F)F)C1=CC=C(Cl)C=C1

Specifications

Melting Point 62°C
Color White
Quantity 5 g
Formula Weight 250.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.