accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOOClClClClCl

CAS RN 18942-25-1

IUPAC Name: tert-butyl 2,3,4,5,6-pentachlorophenyl carbonate
Synonyms: tert-butyl 2,3,4,5,6-pentachlorophenyl carbonate
Molecular Weight (g/mol): 366.44
Molecular Formula: C11H9Cl5O3
InChi Key: GHBRKMLMVFKZKW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)OC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results