Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Pentachlorophenyl Carbonate 95.0+%, TCI America™
SDP

Catalog No. B085910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B085910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B085910G Supplier TCI America Supplier No. B085910G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18942-25-1
Molecular Formula C11H9Cl5O3
Molecular Weight (g/mol) 366.44
MDL Number MFCD00015947
InChI Key GHBRKMLMVFKZKW-UHFFFAOYSA-N
Synonym Boc-O-PCP, Pentachlorophenylcarbonic Acid tert-Butyl Ester
PubChem CID 545481
IUPAC Name tert-butyl (2,3,4,5,6-pentachlorophenyl) carbonate
SMILES CC(C)(C)OC(=O)OC1=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 117°C
Quantity 10 g
Formula Weight 366.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl Pentachlorophenyl Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.