accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18942-46-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Molecular Weight (g/mol): 341.42
Molecular Formula: C16H23NO5S
InChi Key: VRTXRNJMNFVTOM-UHFFFAOYNA-N
SMILES: COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)