Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-S-(4-methoxybenzyl)-L-cysteine, 98%, Thermo Scientific™

Catalog No. AAH6380814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6380814 25 g
AAH6380806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6380814 Supplier Thermo Scientific Chemicals Supplier No. H6380814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18942-46-6
Molecular Formula C16H23NO5S
Molecular Weight (g/mol) 341.42
MDL Number MFCD00038254
InChI Key VRTXRNJMNFVTOM-UHFFFAOYNA-N
Synonym boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine
PubChem CID 87863
IUPAC Name (2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Quantity 25 g
Formula Weight 341.42
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-S-(4-methoxybenzyl)-L-cysteine
EINECSNumber 242-695-5
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.