accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOOO

CAS RN 18959-30-3

IUPAC Name: 5,6-difluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5,6-difluoro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 184.1
Molecular Formula: C8H2F2O3
InChi Key: UATBWQQFLABWKR-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results