Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Difluorophthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. D47165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47165G 5 g
D47161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47165G Supplier TCI America Supplier No. D47165G

Chemical Identifiers

CAS 18959-30-3
Molecular Formula C8H2F2O3
Molecular Weight (g/mol) 184.098
MDL Number MFCD08695033
InChI Key UATBWQQFLABWKR-UHFFFAOYSA-N
PubChem CID 639745
IUPAC Name 5,6-difluoro-2-benzofuran-1,3-dione
SMILES C1=C2C(=CC(=C1F)F)C(=O)OC2=O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 184.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Difluorophthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.