CAS RN 18962-61-3
IUPAC Name:
magnesium(2+) dihydrogen bis((2S)-2-aminobutanedioate)
Synonyms:
magnesium(2+) dihydrogen bis(L-aspartate(2-))
Molecular Weight (g/mol):
288.5
Molecular Formula:
C8H12MgN2O8
InChi Key:
RXMQCXCANMAVIO-CEOVSRFSSA-L
SMILES:
[H+].[H+].[Mg++].N[C@@H](CC([O-])=O)C([O-])=O.N[C@@H](CC([O-])=O)C([O-])=O
1
–
1
of
1
results
1
L-Aspartate de magnesium salt, MP Biomedicals™
CAS: 18962-61-3 Molecular Formula: C8H10MgN2O8-2 Molecular Weight (g/mol): 286.479 InChI Key: XBYSRGUYNYEVEJ-UHFFFAOYSA-L Synonym: l-aspartic acid magnesium salt PubChem CID: 131850428 IUPAC Name: magnesium;2-azanidylbutanedioate;hydron SMILES: [H+].[H+].C(C(C(=O)[O-])[NH-])C(=O)[O-].C(C(C(=O)[O-])[NH-])C(=O)[O-].[Mg+2]