Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartate de magnesium salt, MP Biomedicals™

Catalog No. ICN15040691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15040691 1 kg
ICN15040625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15040691 Supplier MP Biomedicals, Inc Supplier No. 0215040691
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High-purity magnesium salt of L-aspartic acid for research or manufacturing applications.

  • CAS Number: 18962-61-3
  • Molecular Formula: C8H12N2O8Mg
  • Molecular Weight: 288.5 (anhydrous)
  • EC Number: 242-703-7
  • MDL Number: MFCD00044935
  • Optical Rotation: +20°
  • pH: 6.5
  • Solubility: Freely soluble in water
  • Hazard Statement: H317
  • Usage: For research or further manufacturing use only; not for direct human use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18962-61-3
Molecular Formula C8H10MgN2O8-2
Molecular Weight (g/mol) 286.479
InChI Key XBYSRGUYNYEVEJ-UHFFFAOYSA-L
Synonym l-aspartic acid magnesium salt
PubChem CID 131850428
IUPAC Name magnesium;2-azanidylbutanedioate;hydron
SMILES [H+].[H+].C(C(C(=O)[O-])[NH-])C(=O)[O-].C(C(C(=O)[O-])[NH-])C(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 1 kg
Formula Weight 288.5 (Anhydrous)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartate de magnesium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.