CAS RN 189689-94-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PAF Receptor Antagonist, ABT-491
The PAF Receptor Antagonist, ABT-491, also referenced under CAS 189689-94-9, controls the biological activity of PAF Receptor. This small molecule/inhibitor is primarily used for Membrane applications.