accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNClNCl

CAS RN 1897-45-6

IUPAC Name: tetrachlorobenzene-1,3-dicarbonitrile
Synonyms: chlorothalonil m-tcpn daconil meta-TCPN m-tetrachlorophthalonitrile 1,3-dicyanotetrachlorobenzene
Molecular Weight (g/mol): 265.9
Molecular Formula: C8Cl4N2
InChi Key: CRQQGFGUEAVUIL-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(C#N)=C(Cl)C(C#N)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results