Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroisophthalonitrile 98.0+%, TCI America™
SDP

Catalog No. T089525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T089525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T089525G Supplier TCI America Supplier No. T089525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1897-45-6
Molecular Formula C8Cl4N2
Molecular Weight (g/mol) 265.902
MDL Number MFCD00045594
InChI Key CRQQGFGUEAVUIL-UHFFFAOYSA-N
Synonym Perchloroisophthalonitrile, Chlorothalonil
PubChem CID 15910
ChEBI CHEBI:3639
IUPAC Name 2,4,5,6-tetrachlorobenzene-1,3-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1Cl)Cl)Cl)C#N)Cl

Specifications

Physical Form Powder/Crystals
UN Number 3439
Color White/Yellow
Melting Point 251°C
Boiling Point 350°C
Quantity 25 g
Formula Weight 265.90
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrachloroisophthalonitrile
EINECSNumber (3)-1805
RTECSNumber NT2600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.