accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OFFFFFFMgSi

CAS RN 18972-56-0

IUPAC Name: silicon(4+) magnesium(2+) hexahydrate hexafluoride
Synonyms: silicon(4+) magnesium(2+) hexahydrate hexafluoride
Molecular Weight (g/mol): 274.47
Molecular Formula: F6H12MgO6Si
InChi Key: QNZBQSBPLQILSY-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.[F-].[F-].[F-].[F-].[F-].[F-].[Mg++].[Si+4]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results