accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CH3CCH3(Z)(E)(Z)(E)

CAS RN 1898-13-1

IUPAC Name: (1E,3Z,6E,10Z)-3,7,11-trimethyl-14-(propan-2-yl)cyclotetradeca-1,3,6,10-tetraene
Synonyms: (1E,3Z,6E,10Z)-14-isopropyl-3,7,11-trimethylcyclotetradeca-1,3,6,10-tetraene
Molecular Weight (g/mol): 272.48
Molecular Formula: C20H32
InChi Key: DMHADBQKVWXPPM-GMVDCNKVNA-N
SMILES: CC(C)C1CC\C(C)=C/CC\C(C)=C\C\C=C(\C)/C=C/1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)