Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cembrene, 99%, Thermo Scientific™

Catalog No. AC290170025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290170025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290170025 Supplier Thermo Scientific Chemicals Supplier No. 290170025

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1898-13-1
Molecular Formula C20H32
Molecular Weight (g/mol) 272.476
InChI Key DMHADBQKVWXPPM-CZIVKHGLSA-N
Synonym cembrene
PubChem CID 134128866
IUPAC Name (3Z,6Z,10Z)-3,7,11-trimethyl-14-propan-2-ylcyclotetradeca-1,3,6,10-tetraene
SMILES CC1=CCCC(=CCC=C(C=CC(CC1)C(C)C)C)C

Specifications

Melting Point 59°C to 61°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 2.5 g
Specific Rotation +237° to +239° (20°C, 589nm) (c=1, CHCl3)
Formula Weight 272.46
Percent Purity ≥98.5%
Physical Form Fine Needle-like Crystalline Powder
Chemical Name or Material Cembrene, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.