CAS RN 1898-72-2
IUPAC Name:
2,4-dimethoxy-1,3,5-triazine
Synonyms:
2,4-dimethoxy-1,3,5-triazine
Molecular Weight (g/mol):
141.13
Molecular Formula:
C5H7N3O2
InChi Key:
RIRBAVAYPRSMRH-UHFFFAOYSA-N
SMILES:
COC1=NC(OC)=NC=N1
1
–
1
of
1
results
1
2,4-Dimethoxy-1,3,5-triazine 98.0+%, TCI America™
CAS: 1898-72-2 Molecular Formula: C5H7N3O2 Molecular Weight (g/mol): 141.13 InChI Key: RIRBAVAYPRSMRH-UHFFFAOYSA-N PubChem CID: 243963 IUPAC Name: 2,4-dimethoxy-1,3,5-triazine SMILES: COC1=NC(=NC=N1)OC