accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18981-98-1

IUPAC Name: (heptyloxy)sulfonic acid sodium
Synonyms: heptyloxysulfonic acid sodium
Molecular Weight (g/mol): 219.25
Molecular Formula: C7H16NaO4S
InChi Key: SLGMFWJQQBPOMZ-UHFFFAOYSA-N
SMILES: [Na].CCCCCCCOS(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results