accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCH3NH3CCH3

CAS RN 1899-02-1

IUPAC Name: N,N,N-trimethylanilinium hydroxide
Synonyms: trimethylphenylammonium hydroxide
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: HADKRTWCOYPCPH-UHFFFAOYSA-M
SMILES: [OH-].C[N+](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)