accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18995-35-2

IUPAC Name: 1-(tert-butoxy)-4-chlorobenzene
Synonyms: 1-(tert-butoxy)-4-chlorobenzene
Molecular Weight (g/mol): 184.66
Molecular Formula: C10H13ClO
InChi Key: NEJWTQIEQDHWTR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results