Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxy-4-chlorobenzene 95.0+%, TCI America™
SDP

Catalog No. B227825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B227825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B227825G Supplier TCI America Supplier No. B227825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18995-35-2
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.663
MDL Number MFCD04038411
InChI Key NEJWTQIEQDHWTR-UHFFFAOYSA-N
Synonym tert-Butyl 4-Chlorophenyl Ether
PubChem CID 140458
IUPAC Name 1-chloro-4-[(2-methylpropan-2-yl)oxy]benzene
SMILES CC(C)(C)OC1=CC=C(C=C1)Cl

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
Formula Weight 184.66
Percent Purity ≥95.0% (GC)
Chemical Name or Material 1-tert-Butoxy-4-chlorobenzene
EINECSNumber (3)-0845
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.