CAS RN 19014-29-0
IUPAC Name:
{4-[4-(dihydroxyboranyl)phenoxy]phenyl}boronic acid
Synonyms:
4-[4-(dihydroxyboranyl)phenoxy]phenylboronic acid
Molecular Weight (g/mol):
257.84
Molecular Formula:
C12H12B2O5
InChi Key:
DFPCWEXYGPRULG-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(OC2=CC=C(C=C2)B(O)O)C=C1
1
–
1
of
1
results
1
4,4'-Oxybis(1,4-phenylene)diboronic acid, 97%, Thermo Scientific™
CAS: 19014-29-0 Molecular Formula: C12H12B2O5 Molecular Weight (g/mol): 257.84 MDL Number: MFCD00433207 InChI Key: DFPCWEXYGPRULG-UHFFFAOYSA-N Synonym: oxybis 4,1-phenylene diboronic acid,4,4'-oxybis 1,4-benzene diboronic acid,4,4'-oxybis 1,4-phenylene diboronic acid,4-4-boronophenoxy phenyl boronic acid,4-4-dihydroxyboranyl phenoxy phenylboronic acid,oxybis 4,1-phenylene bisboronic acid,oxydi 4,1-phenylene diboronic acid PubChem CID: 2832120 IUPAC Name: [4-(4-boronophenoxy)phenyl]boronic acid SMILES: OB(O)C1=CC=C(OC2=CC=C(C=C2)B(O)O)C=C1