Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Oxybis(1,4-phenylene)diboronic acid, 97%, Thermo Scientific™

Catalog No. AAH53158MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53158MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53158MD Supplier Thermo Scientific Chemicals Supplier No. H53158MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19014-29-0
Molecular Formula C12H12B2O5
Molecular Weight (g/mol) 257.84
MDL Number MFCD00433207
InChI Key DFPCWEXYGPRULG-UHFFFAOYSA-N
Synonym oxybis 4,1-phenylene diboronic acid, 4,4'-oxybis 1,4-benzene diboronic acid, 4,4'-oxybis 1,4-phenylene diboronic acid, 4-4-boronophenoxy phenyl boronic acid, 4-4-dihydroxyboranyl phenoxy phenylboronic acid, oxybis 4,1-phenylene bisboronic acid, oxydi 4,1-phenylene diboronic acid
PubChem CID 2832120
IUPAC Name [4-(4-boronophenoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OC2=CC=C(C=C2)B(O)O)C=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 257.84
Percent Purity 97%
Chemical Name or Material 4,4'-Oxybis(1,4-phenylene)diboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.