CAS RN 19044-88-3
IUPAC Name:
4-(dipropylamino)-3,5-dinitrobenzene-1-sulfonamide
Synonyms:
oryzalin
Molecular Weight (g/mol):
346.36
Molecular Formula:
C12H18N4O6S
InChi Key:
UNAHYJYOSSSJHH-UHFFFAOYSA-N
SMILES:
CCCN(CCC)C1=C(C=C(C=C1[N+]([O-])=O)S(N)(=O)=O)[N+]([O-])=O
1
–
1
of
1
results
1
Oryzalin, SPEX CertiPrep™
CAS: 19044-88-3 Molecular Formula: C12H18N4O6S Molecular Weight (g/mol): 346.358 InChI Key: UNAHYJYOSSSJHH-UHFFFAOYSA-N PubChem CID: 29393 ChEBI: CHEBI:73163 IUPAC Name: 4-(dipropylamino)-3,5-dinitrobenzenesulfonamide SMILES: CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])S(=O)(=O)N)[N+](=O)[O-]