accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3O

CAS RN 1906-82-7

IUPAC Name: ethyl 2-acetamidoacetate
Synonyms: ethyl 2-acetamidoacetate
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: AMBDTBHJFINMSE-UHFFFAOYSA-N
SMILES: CCOC(=O)CNC(C)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)