Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetamidoacetate 98%, Thermo Scientific™

Catalog No. AC117951000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC117951000 100 g
AC117950250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117951000 Supplier Thermo Scientific Chemicals Supplier No. 117951000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1906-82-7
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
InChI Key AMBDTBHJFINMSE-UHFFFAOYSA-N
Synonym ethyl acetamidoacetate, ac-gly-oet, n-acetylglycine ethyl ester, ethyl n-acetylglycinate, ethylaceturate, ethyl acetaminoacetate, ethyl 2-acetylamino acetate, glycine, n-acetyl-, ethyl ester, acetylglycine ethyl ester, acetyl-glycine ethyl ester
PubChem CID 74702
IUPAC Name ethyl 2-acetamidoacetate
SMILES CCOC(=O)CNC(=O)C

Specifications

Melting Point 42°C to 47°C
Color White
Infrared Spectrum Authentic
Linear Formula CH3CONHCH2CO2C2H5
Quantity 100 g
Beilstein 04, 354
Formula Weight 145.16
Percent Purity ≥97.5%
Physical Form Shiny Crystalline Powder
Chemical Name or Material Ethyl acetamidoacetate, 98%
EINECSNumber 217-608-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.