accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3NOH3C

CAS RN 19060-15-2

IUPAC Name: 4,4-dimethoxybutan-1-aminium
Synonyms: 4,4-dimethoxybutan-1-aminium
Molecular Weight (g/mol): 134.2
Molecular Formula: C6H16NO2
InChi Key: TYVAXMOICMBSMT-UHFFFAOYSA-O
SMILES: COC(CCC[NH3+])OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)