CAS RN 190654-01-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cdk1 Inhibitor, CGP74514A
The Cdk1 Inhibitor, CGP74514A, also referenced under CAS 190654-01-4, controls the biological activity of Cdk1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.