accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH3C

CAS RN 190974-21-1

IUPAC Name: N9,N10-bis(4-methylphenyl)-N9,N10-diphenylanthracene-9,10-diamine
Synonyms: N9,N10-bis(4-methylphenyl)-N9,N10-diphenylanthracene-9,10-diamine
Molecular Weight (g/mol): 540.71
Molecular Formula: C40H32N2
InChi Key: HTJPPQKJCPTAED-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C2C=CC=CC2=C(N(C2=CC=CC=C2)C2=CC=C(C)C=C2)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results