CAS RN 191090-32-1
IUPAC Name:
5,5-diphenyl-4-(propan-2-yl)-1,3-oxazolidin-2-one
Synonyms:
4-isopropyl-5,5-diphenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
281.36
Molecular Formula:
C18H19NO2
InChi Key:
PHTOJBANGYSTOH-UHFFFAOYNA-N
SMILES:
CC(C)C1NC(=O)OC1(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98.0+%, TCI America™
CAS: 191090-32-1 Molecular Formula: C18H19NO2 Molecular Weight (g/mol): 281.355 MDL Number: MFCD03093555 InChI Key: PHTOJBANGYSTOH-MRXNPFEDSA-N PubChem CID: 11300512 IUPAC Name: (4R)-5,5-diphenyl-4-propan-2-yl-1,3-oxazolidin-2-one SMILES: CC(C)C1C(OC(=O)N1)(C2=CC=CC=C2)C3=CC=CC=C3