Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98.0+%, TCI America™
SDP

Catalog No. I07615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07615G 5 g
I07611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07615G Supplier TCI America Supplier No. I07615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 191090-32-1
Molecular Formula C18H19NO2
Molecular Weight (g/mol) 281.355
MDL Number MFCD03093555
InChI Key PHTOJBANGYSTOH-MRXNPFEDSA-N
PubChem CID 11300512
IUPAC Name (4R)-5,5-diphenyl-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1C(OC(=O)N1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 254°C
Color White
Quantity 5 g
Formula Weight 281.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.