accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19111-87-6

IUPAC Name: 2-bromotriphenylene
Synonyms: 2-bromotriphenylene
Molecular Weight (g/mol): 307.19
Molecular Formula: C18H11Br
InChi Key: GEDOYYDMCZUHNW-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results