Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromotriphenylene 98.0+%, TCI America™
SDP

Catalog No. B44425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44425G 5 g
B44421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44425G Supplier TCI America Supplier No. B44425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19111-87-6
Molecular Formula C18H11Br
Molecular Weight (g/mol) 307.19
MDL Number MFCD16659043
InChI Key GEDOYYDMCZUHNW-UHFFFAOYSA-N
PubChem CID 18381174
IUPAC Name 2-bromotriphenylene
SMILES BrC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 5 g
Formula Weight 307.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromotriphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.