accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19117-31-8

IUPAC Name: tert-butyl(phenylsulfanyl)amine
Synonyms: tert-butyl(phenylsulfanyl)amine
Molecular Weight (g/mol): 181.3
Molecular Formula: C10H15NS
InChi Key: AAQBFMPKCDTAJD-UHFFFAOYSA-N
SMILES: CC(C)(C)NSC1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results