Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylbenzenesulfenamide 96.0+%, TCI America™
SDP

Catalog No. B22405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22405G 5 g
B22401G 1 g
B224025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22405G Supplier TCI America Supplier No. B22405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19117-31-8
Molecular Formula C10H15NS
Molecular Weight (g/mol) 181.30
MDL Number MFCD03844776
InChI Key AAQBFMPKCDTAJD-UHFFFAOYSA-N
Synonym N-tert-Butylphenylsulfenamide
PubChem CID 11182990
IUPAC Name tert-butyl(phenylsulfanyl)amine
SMILES CC(C)(C)NSC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 181.30
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-tert-Butylbenzenesulfenamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.