accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNClNNHH3CCH3N

CAS RN 1912-24-9

IUPAC Name: 6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: atrazine
Molecular Weight (g/mol): 215.69
Molecular Formula: C8H14ClN5
InChi Key: MXWJVTOOROXGIU-UHFFFAOYSA-N
SMILES: CCNC1=NC(Cl)=NC(NC(C)C)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)