accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1913-72-0

H3CCH3ONNHN

CAS RN 1913-72-0

IUPAC Name: 5,7-dimethyl-1H,4H-pyrido[2,3-d]pyrimidin-4-one
Synonyms: 5,7-dimethyl-1H-pyrido[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: HWGAGILTAOQBPD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C(=O)N=CNC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results