Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine, 98%, Thermo Scientific™

Catalog No. AC271910010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC271910010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC271910010 Supplier Thermo Scientific Chemicals Supplier No. 271910010

missing translation for 'validMainframeMsg'

Specifications

CAS 1913-72-0
CAS Min % 97.5
CAS Max % 100.0
Color Beige
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H9N3O
MDL Number MFCD00138563
Quantity 1 g
Synonym 5,7-dimethylpyrido 2,3-d pyrimidin-4 3h-one, 5,7-dimethyl-1h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-3-hydropyridino 2,3-d pyrimidin-4-one, 5,7-dimethyl-3h-pyrido 2,3-d pyrimidin-4-one, 5,7-dimethyl-4-hydroxypyrido 2,3-d pyrimidine, 5?7-dimethyl-4-hydroxypyrido 2?3-d pyrimidine, 5,7-dimethyl-3h,4h-pyrido 2,3-d pyrimidin-4-one, pyrido 2,3-d pyrimidin-4 3h-one,5,7-dimethyl, 5,7-dimethyl-4-oxo-3,4-dihydropyrido 2,3-d pyrimidine
InChI Key HWGAGILTAOQBPD-UHFFFAOYSA-N
SMILES CC1=CC(C)=C2C(=O)N=CNC2=N1
IUPAC Name 5,7-dimethyl-1H-pyrido[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol) 175.19
PubChem CID 5394613
Formula Weight 175.19
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material 5,7-Dimethyl-4-hydroxypyrido[2,3-d]pyrimidine
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.