accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHOOOOCH3OHO

CAS RN 191605-10-4

IUPAC Name: 2,3-bis(4-methoxybenzoyloxy)butanedioic acid
Synonyms: 2,3-bis(4-methoxybenzoyloxy)butanedioic acid
Molecular Weight (g/mol): 418.35
Molecular Formula: C20H18O10
InChi Key: KWWCVCFQHGKOMI-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(OC)C=C1)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results